Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naphthalen-1-yl 3,5-dinitrobenzoate | 93261-39-3 | MFCD00376091 | 99.5% | 338.27 g/mol | C17H10N2O6 | 25MG
SDP

Supplier:  Medchemexpress LLC HY12597525MG

Encompass_Preferred

1-Naphthyl 3,5-dinitrobenzoate (CAS 93261-39-3) is a small-molecule research compound used in biochemical studies as a 5-lipoxygenase inhibitor and mPGES-1 modulator. It is reported with low-micromolar inhibitory activity in enzymatic and whole-blood assays and is supplied as a high-purity solid for in vitro research applications.

  • High purity solid suitable for in vitro research.
  • Molecular formula C17H10N2O6.
  • Molecular weight 338.27 g/mol.
  • Reported 5-LOX IC50 of 1.04 μM.
  • Additional activity: mPGES-1 IC50 3.6 μM; human whole blood IC50 8.6 μM.
  • Commonly supplied in small-mass packaging such as 25 MG.

Catalog No. 50-002-37818


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.