Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Naphthalene-1-boronic acid, 13922-41-3, MFCD00019722, 25g

Supplier:  Matrix Scientific 00461425G

Encompass_Preferred

Molecular Formula C10H9BO2, Purity 97%, Molecular Weight 171.99, Melting Point 202°

Catalog No. 50-011-7008


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.