Learn More
Medchemexpress LLC Naphthalene-1-carboxamide | 890128-81-1 | MFCD30534392 | 96.7% | 439.39 g/mol | C23H16F3N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10041910MG
Encompass_Preferred
BFH772 is a potent, orally active small-molecule inhibitor of vascular endothelial growth factor receptor 2 (VEGFR2). It inhibits VEGFR2 kinase with an IC50 of 3 nM and is supplied as a solid research compound for biochemical and cellular studies of angiogenesis signaling.
  • Potent VEGFR2 inhibition (IC50 3 nM).
  • Orally active small-molecule suitable for in vitro and preclinical studies.
  • High purity solid appropriate for biochemical assays and formulation work.
  • Molecular weight 439.39 g/mol and CAS 890128-81-1 for unambiguous identification.
  • Available in small research pack sizes for screening and characterization.
Catalog No. 50-002-14793