Learn More
Medchemexpress LLC Naphthalene, 2-(1-methylethyl)- | 2027-17-0 | MFCD00014322 | 95.3% | 170.25 g/mol | C13H14 | 5 G

Supplier: Medchemexpress LLC HYW1501665G
2-Isopropylnaphthalene is a biochemical assay reagent provided for laboratory research use only. It is used in biochemical and analytical applications and is characterized by its chemical identity and reported purity suitable for routine laboratory handling.
- Intended for research use only, not for human, clinical, or diagnostic applications.
- Colorless to light yellow liquid suitable for analytical procedures.
- Purity reported at approximately 95.3% for typical batches.
- Contains CAS number 2027-17-0 for unambiguous identification.
- Available in small lab-scale packaging for convenient handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.