Learn More
Medchemexpress LLC Naphthalene, 2-ethenyl- | 827-54-3 | MFCD00004125 | 99.8% | 154.21 g/mol | C12H10 | 25 G

Supplier: Medchemexpress LLC 25GHYW015553
2-Vinylnaphthalene (IUPAC name: naphthalene, 2-ethenyl-) is an aromatic vinyl monomer used as a building block in polymer synthesis and materials research. It is supplied as a white to off-white solid with high chemical purity for research applications.
- High purity: 99.79% (analytical grade)
- Molecular formula C12H10; molecular weight 154.21 g/mol
- Soluble in DMSO; typical solubility ~100 mg/mL with sonication
- Suitable for polymerization and organic synthesis applications
- Stable when stored sealed, away from moisture; follow recommended storage conditions for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.