Learn More
Medchemexpress LLC Naphthazarin (Standard) | 475-38-7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN7526
Encompass_Preferred
Naphthazarin (DHNQ) is a naturally occurring compound that triggers apoptosis and exhibits anti-tumor effects. It works through various cellular mechanisms including oxidative stress, activation of mitochondrial apoptosis-inducing factor (AIF), depolymerization of microtubules, interference with lysosomal function, and p53-dependent p21 activation.
  • Effective through various cellular mechanisms
  • Triggers apoptosis
  • Exhibits anti-tumor effects
  • Analytical standard for reference assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-20652