Learn More
Apexbio Technology LLC β-Naphthoflavone 6051-87-2 200mg
SDP
Supplier:  Apexbio Technology LLC A4752200
Encompass_Preferred
-Naphthoflavone (CAS 6051-87-2) also known as 5 6-benzoflavone is a small molecule that acts as a potent agonist of the aryl hydrocarbon receptor (AhR) Activation of AhR by -naphthoflavone upregulates the expression of metabolizing enzymes including multiple cytochrome P450 isoforms (CYPs) and uridine 5 -diphospho-glucuronosyltransferases (UGTs) This compound is widely utilized in biomedical research to investigate xenobiotic metabolism enzyme induction and the regulation of detoxification pathways Additionally it has been studied for its potential role in chemoprevention due to its modulatory effects on metabolic enzymes
Catalog No. 50-000-96955