Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0164100MG
Encompass_Preferred
Naphthol AS-BI is a substrate of β-glucuronidase and produces a bright red effect after staining biological tissues.
  • For research use only.
  • Not for sale to patients.
Catalog No. 50-004-10522