Learn More
Medchemexpress LLC HY-104068 100mg Medchemexpress, Naphthol AS-E CAS:92-78-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104068100MG
Encompass_Preferred
Medchemexpress, HY-104068 100mg Naphthol AS-E CAS:92-78-4 Naphthol AS-E is a potent and cell-permeable inhibitor of KIX-KID interaction. Naphthol AS-E directly binds to the KIX domain of CBP (Kd:8.6 µM), blocks the interaction between the KIX domain and the KID domain of CREB with IC50 of 2.26 µM. Naphthol AS-E can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0679