Learn More
Medchemexpress LLC Naphthol AS-E | 92-78-4 | MFCD00021639 | 98.0% | 297.74 g/mol | C17H12ClNO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY104068500MG
Encompass_Preferred
Naphthol AS-E is a small-molecule, cell-permeable inhibitor that binds the KIX domain of CREB-binding protein (CBP) and disrupts the KIX-KID interaction. It is used in biochemical and cell-based studies of transcriptional regulation and cancer research, with reported binding affinity (Kd ≈ 8.6 μM) and inhibitory activity (IC50 ≈ 2.26 μM).
  • Cell-permeable inhibitor of KIX-KID interaction.
  • Directly binds the KIX domain of CBP.
  • Useful for biochemical and cell-based assays.
  • High reported purity suitable for research applications.
  • Available in solid and solution formats for flexible use.
Catalog No. 50-002-41726