Learn More
AdipoGen Naphthol AS-MX phosphate
SDP
Supplier:  AdipoGen CDXN0127G001
Encompass_Preferred
Chemical. CAS 1596-56-1. Formula C19H18NO5P. MW BD9837. Naphthol AS-MX Phosphate is a fluorogenic substrate for alkaline phosphatases. Naphthol AS-MX Phosphate is converted to naphthol AS-MX displaying excitation/emission spectra of 388/512nm. The fluorescence is a quantitative marker of acid and alkaline phosphatase activity. Acid and alkaline phosphatases are often used as clinical markers of disease, since the concentration of human acid and alkaline phosphatases undergo pronounced changes in particular diseases, resulting in unusually high or low concentrations. It is a histochemical substrate commonly used in conjunction with diazonium salts to form a specific azo dye for immunohistology, immunoblotting and dot blot applications.
Catalog No. 50-256-0400