Learn More
Apexbio Technology LLC Naphthol AS-MX phosphate 1596-56-1 100mg
SDP
Supplier:  Apexbio Technology LLC A9086100
Encompass_Preferred
Naphthol AS-MX phosphate (CAS 1596-56-1) is a small molecule inhibitor that disrupts the interaction between cAMP response element-binding protein (CREB) and CREB-binding protein (CBP) key components of a transcription factor complex In lung cancer cells it demonstrates antiproliferative effects as evidenced by an IC50 of 3 701 mol/L and inhibits both colony formation and anchorage-independent growth Naphthol AS-MX phosphate is utilized in studies investigating KRAS-mutant lung cancers particularly in the context of chemoresistance and poor prognosis
Catalog No. 50-000-96256