Learn More
Medchemexpress LLC Naphthol AS-MX phosphate | 1596-56-1 | MFCD00042711 | 99.5% | 371.32 g/mol | C19H18NO5P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY104067100MG
Encompass_Preferred
Naphthol AS-MX phosphate is the phosphate salt of Naphthol AS-MX supplied as a purified solid for research use. It acts as a CREB-CBP complex inhibitor and is used in biochemical and cell biology studies, including enzyme inhibition assays and cell proliferation experiments.
  • High purity (99.5%) solid suitable for research applications.
  • Available in small-scale package sizes for laboratory testing.
  • Soluble in DMSO at 125 mg/mL for convenient stock preparation.
  • Stable when stored sealed and protected from moisture; follow recommended storage conditions.
  • Compatible with biochemical assays as an inhibitor and with enzyme substrate applications.
Catalog No. 50-002-42819