Learn More
Medchemexpress LLC α-Naphtholbenzein | 145-50-6 | MFCD00078492 | 87.2% | 374.44 | C27H18O2 | 5G

Supplier: Medchemexpress LLC HY1254575G
α-Naphtholbenzein is a synthetic dye used as a colorimetric pH indicator and analytical reagent. It is supplied as a solid powder intended for dissolution in organic solvents for research and laboratory applications.
- Used as a pH indicator and colorimetric reagent.
- Supplied as a brown to reddish brown solid powder.
- Purity 87.2%.
- Molecular formula C27H18O2; molecular weight 374.44 g/mol.
- Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required.
- Store powder at -20°C (stable for 3 years) or 4°C (stable for 2 years); store in solvent at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.