Learn More
Medchemexpress LLC β-Naphthoxyethanol | 93-20-9 | MFCD00016809 | 98.1% | 188.22 | C12H12O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW016083
Encompass_Preferred
Anavenol (β-naphthoxyethanol, CAS 93-20-9) is a small-molecule analgesic historically used in veterinary contexts and supplied for research and reagent use. It is a white to off-white solid with formula C12H12O2 and molecular weight ~188.22, reported at high purity. Typical handling involves dissolution in DMSO for assays and formulation.
  • High reported purity (~98.1%).
  • Solid, white to off-white appearance for easy visual identification.
  • Suitable for veterinary analgesic research and related pharmacology studies.
  • Soluble in DMSO for preparation of stock solutions and assays.
  • Available in multiple small pack sizes to support laboratory experiments.
Catalog No. 50-003-02362