Learn More
Electron Microscopy Sciences Naphtol Green B Stain 1% 250 ML
SDP
Supplier:  Electron Microscopy Sciences 2610903
Encompass_Preferred
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.
Catalog No. 50-316-62