Learn More
Aniara Diagnostica LLC NAPI 0.02M PDR PH 7.0 1L 10 EA
Supplier:  Aniara Diagnostica LLC A12952710
Encompass
Medicago NaPi 0.02 M Buffer pH 7.0 supplied as preweighed powder in sealed pouch yields 1000 mL of 0.02 M Sodium Phosphate buffer with pH 7.0 at 25C. Used for biochemical and molecular biology applications. 10 pouches.
Catalog No. 50-100-8864