Learn More
Selleck Chemical LLC Naporafenib S8745-1G
SDP
Supplier:  Selleck Chemical LLC S87451G
Encompass_Preferred
Naporafenib (LXH254) is a type II ATP-competitive inhibitor that inhibits both B- and CRAF kinase activities at picomolar concentrations with a high degree of selectivity against a panel of 456 human kinases and in cell-based assays
Catalog No. 50-000-24087