Learn More
Medchemexpress LLC Napropamide | 15299-99-7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB197210MM/1ML
Encompass_Preferred
Napropamide is an amide herbicide that controls weeds in fruits, vegetables, tobacco, and ornamental plants by inhibiting DNA synthesis and cell division. It exhibits moderate to high persistence with a half-life of 24 to 131 days and is susceptible to photodegradation. Studies indicate low acute toxicity, no genotoxicity, carcinogenicity, neurotoxicity, or adverse effects on fertility, reproduction, embryotoxicity, or teratogenicity.
  • Amide herbicide
  • Controls weeds in fruits, vegetables, tobacco, and ornamental plants
  • Inhibits DNA synthesis and cell division
  • Moderate to high persistence in field
  • Low acute toxicity
  • No genotoxicity, carcinogenicity, or neurotoxicity
  • No adverse effects on fertility, reproduction, embryotoxicity, or teratogenicity
Catalog No. 50-004-11105