Learn More
Medchemexpress LLC Naproxen | 22204-53-1 | C14H14O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1503010MM/1ML
Encompass_Preferred
Naproxen is a COX-1 and COX-2 inhibitor, supplied as a 10mM solution in 1mL. It is an equipotent inhibitor of COX-1 and COX-2 in intact cells. Naproxen exerts anti-inflammatory and antifibrotic effects in a mouse model of bleomycin-induced lung fibrosis and downregulates TGF-β levels and Smad3/4 complex formation. It also inhibits the time-courses of pain, fever, and PGE2.
  • COX-1 and COX-2 inhibitor
  • Supplied as a 10 mM solution in 1 mL
  • Exerts anti-inflammatory and antifibrotic effects
  • Downregulates TGF-β levels
  • Inhibits pain, fever, and PGE2
Catalog No. 50-003-75484