Learn More
Medchemexpress LLC Naproxen | 22204-53-1 | MFCD00078864 | C14H14O3 | 25 G

Supplier: Medchemexpress LLC 25GHY15029
Naproxen is a non-steroidal anti-inflammatory drug (NSAID) that acts as a COX-1 and COX-2 inhibitor, with IC50s of 8.72 and 5.15 μM respectively in cell assays. It is for research use only.
- Inhibits COX-1 and COX-2 enzymes
- High purity of 99.98%
- Target: COX and Autophagy
- Pathway: Immunology/Inflammation and Autophagy
- Stable for 3 years at room temperature
- Stable in solvent for 2 years at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.