Learn More
Medchemexpress LLC (±)-Naproxen | 23981-80-8 | MFCD00439456 | C14H14O3 | 10 G

Supplier: Medchemexpress LLC HY1502910G
(±)-Naproxen, also known as (Rac)-Naproxen, is a racemate of Naproxen. It functions as an inhibitor for both COX-1 and COX-2 with IC50s of 8.72 μM and 5.15 μM respectively. This compound has a molecular formula of C14H14O3 and a molecular weight of 230.26, presenting as a white to light yellow solid. It is intended for research use only.
- Inhibits COX-1 and COX-2
- Molecular weight 230.26
- White to light yellow solid
- Store powder at -20°C for 3 years, 4°C for 2 years
- Store in solvent at -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.