Learn More
Medchemexpress LLC (±)-Naproxen | 23981-80-8 | MFCD00439456 | C14H14O3 | 5 G

Supplier: Medchemexpress LLC HY150295G
(±)-Naproxen is a racemate of Naproxen. It functions as a COX-1 and COX-2 inhibitor, with IC50 values of 8.72 and 5.15 μM, respectively.
- Racemate of naproxen
- Inhibits COX-1 and COX-2
- Purity of 99.79%
- Molecular weight: 230.26
- Solid appearance
- White to light yellow color
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.