Learn More
Cayman Chemical -Naproxen-d3 500ug
SDP
Supplier:  Cayman Chemical 18258500 UG
Encompass_Preferred
An internal standard for the quantification of naproxen by GC- or LC-MS
Catalog No. 50-296-5909