Learn More
Medchemexpress LLC Naproxen etemesil | 385800-16-8 | 99.89% | 336.40 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1967510MM/1ML
Encompass_Preferred
Naproxen etemesil is a lipophilic, non-acidic, inactive proagent of naproxen that is hydrolysed to pharmacologically active Naproxen once absorbed. Naproxen is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay. It is for research use only and not sold to patients.
  • Lipophilic, non-acidic, inactive proagent of naproxen.
  • Hydrolysed to pharmacologically active Naproxen upon absorption.
  • Naproxen is a COX-1 and COX-2 inhibitor.
  • IC50s of 8.72 μM for COX-1 and 5.15 μM for COX-2 in cell assay.
  • Purity: 99.89%.
Catalog No. 50-004-01265