Learn More
Medchemexpress LLC Naproxen etemesil | 385800-16-8 | 99.89% | 336.40 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19675100MG
Encompass_Preferred
Naproxen etemesil is described as a lipophilic, non-acidic, and inactive proagent of naproxen. It becomes pharmacologically active Naproxen after hydrolysis and absorption. Naproxen functions as an inhibitor of both COX-1 and COX-2, with IC50 values of 8.72 μM and 5.15 μM respectively in cell assays.
  • Functions as an inactive proagent that converts to active naproxen upon absorption.
  • Its active form is a COX-1 and COX-2 inhibitor.
  • Has demonstrated anti-inflammatory and antifibrotic effects in a mouse model of bleomycin-induced lung fibrosis.
  • Downregulates TGF-β levels and Smad3/4 complex formation.
  • Inhibits the time-courses of pain, fever, and PGE2.
Catalog No. 50-004-00529