Learn More
Medchemexpress LLC Naproxen etemesil | 385800-16-8 | 99.9% | 336.40 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY196755MG
Encompass_Preferred
Naproxen etemesil is a lipophilic, non-acidic, inactive proagent of naproxen that is hydrolyzed to pharmacologically active naproxen once absorbed. Naproxen is a well-known nonsteroidal anti-inflammatory drug, acting as an equipotent inhibitor of COX-1 and COX-2 in intact cells. This compound offers a targeted approach for anti-inflammatory research due to its conversion within the body.
  • Inactive prodrug of naproxen
  • Hydrolyzed to pharmacologically active naproxen
  • Functions as a COX-1 and COX-2 inhibitor
  • Demonstrates anti-inflammatory and antifibrotic effects
  • Downregulates TGF-β levels
  • Inhibits pain, fever, and PGE2
Catalog No. 50-004-00628