Learn More
Medchemexpress LLC Naproxen etemesil | 385800-16-8 | MFCD30532651 | 99.9% | 336.40 | C17H20O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1967510MG
Encompass_Preferred
Naproxen etemesil is a lipophilic, non-acidic prodrug of naproxen that is hydrolyzed to pharmacologically active naproxen after absorption. It is used in research to study cyclooxygenase (COX) inhibition and related pharmacology. The compound has formula C17H20O5S, molecular weight 336.40 Da, CAS 385800-16-8, and is supplied at high purity for laboratory use.
  • Prodrug converted to naproxen in biological systems.
  • Suitable for cyclooxygenase (COX) inhibition studies.
  • High purity appropriate for research applications.
  • Available in small lab-scale quantities for testing.
  • Chemical formula C17H20O5S; molecular weight 336.40 Da.
Catalog No. 50-002-94778