Learn More
AdipoGen -Naproxen methyl ester
SDP
Supplier:  AdipoGen CDXN0557M050
Encompass_Preferred
Chemical. CAS 26159-35-3. Formula C15H16O3. MW 244.29. The metabolite Naproxen methyl ester Naproxen Impurity E is a degradation product from Naproxen which is a non-steroidal anti-inflammatory drug NSAID used to in the treatment of pain, menstrual cramps, inflammatory diseases such as rheumatoid arthritis, gout and fever. Ibuprofen is an anti-inflammatory COX inhibitor. This product can be used as a control esterase substrate, as a working standard or secondary analytical reference standard or as an organic building block and intermediate for synthesis.
Catalog No. 50-256-0356