Learn More
Selleck Chemical LLC Naproxen Sodium S1626-50mg
SDP
Supplier:  Selleck Chemical LLC S162650MG
Encompass_Preferred
Naproxen Sodium (RS-3650) is a COX inhibitor for COX-1 and COX-2 with IC50 of 8 7 M and 5 2 M respectively
Catalog No. 50-311-0959