Learn More
Genetex Inc Napsin A Monoclonal Antibody [MSVA-555R] HistoMAX™ (GTX04366) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04366
Encompass_Preferred
"The Napsin A antibody [MSVA-555R] HistoMAX™ (GTX04366) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human lung adenocarcinoma (LUAD) tissue using GTX04366 Napsin A antibody [MSVA-112R] HistoMAX. Adenocarcinoma of the lung with strong expression of Napsin A."
Catalog No. NC3663018