Learn More
Sigma Aldrich Fine Chemicals Biosciences NAPTM 5 50 ST Cytiva 17 08
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17085302
Encompass
Disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run.NAP(TM) Columns are disposable columns prepacked with Sephadex(R) G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration washing and sample elution.
Catalog No. 50-290-4886