Learn More
Cayman Chemical NArachidonoyl Taurine
SDP
Supplier:  Cayman Chemical 10005537500 UG
Encompass_Preferred
An arachidonoyl amino acid; an activator of TRPV1 and TRPV4 (EC50s = 28 and 21 µM, respectively); increases calcium flux in HIT-T15 pancreatic β-cells and INS-1 rat islet cells at 10 µM; increases insulin secretion from 832/13 INS-1 pancreatic β-cells; levels are changed in mouse brain following administration of Δ9-THC
Catalog No. 50-289-0302