Learn More
Cayman Chemical NArachidonyl Maleimide
SDP
Supplier:  Cayman Chemical 100075175 MG
Encompass_Preferred
A potent inhibitor of monoacylglycerol lipase (IC50 = 140 nM in rat cerebellar membranes)
Catalog No. 50-289-0809