Learn More
Cayman Chemical Nargencn 5mg
SDP
Supplier:  Cayman Chemical 254565 MG
Encompass_Preferred
A macrolide antibiotic that is selective for S. aureus, MRSA, and M. luteus (MICs = 0.6, 0.3, and 2.5 μg/ml, respectively) over a panel of 11 Gram-positive and Gram-negative bacteria (MICs = >80 μg/ml); inhibits S. aureus DnaE in the presence of DNase I-activated DNA and E. coli DnaE at 0.00001-0.1 and 0.01-100 μg/mL, respectively; inhibits LPS-induced cytokine expression and nitric oxide production in murine BV-2 cells at 1 μM; reduces HL-60 cell proliferation increases cell differentiation when used in combination with 1,25-dihydroxyvitamin D3 or all-trans retinoic acid at 200 μM; reduces the number of CFUs in infected kidneys by 100,000-fold in a murine model of S. aureus infection at 50 mg/kg, p.o
Catalog No. 50-293-6572