Learn More
Medchemexpress LLC Naringenin 7-O-glucoside | 529-55-5 | MFCD00210548 | 99.2% | 434.39 | C21H22O10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN154910MG
Encompass_Preferred
Prunin (Naringenin 7-O-glucoside) is a flavanone glycoside used in biochemical research for antiviral and phosphatase inhibition studies. It is reported to inhibit human enterovirus A71 (HEV-A71) and to inhibit protein tyrosine phosphatase 1B (PTP1B) with an IC50 of 5.5 μM. The compound is supplied as a high-purity analytical standard suitable for in vitro assays.
Catalog No. 50-002-95832