Learn More
Medchemexpress LLC HY-N0804 5mg Medchemexpress, Narirutin CAS:14259-46-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08045MG
Encompass_Preferred
Medchemexpress, HY-N0804 5mg Narirutin CAS:14259-46-2 Narirutin, one of the active constituents isolated from Citrus unshiu, has antioxidant and anti-inflammatory activities. Narirutin is a shikimate kinase inhibitor with anti-tubercular potency[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1405