Learn More
Cayman Chemical Nat-20 S-yn 1mg
SDP
Supplier:  Cayman Chemical 90013691 MG
Encompass_Preferred
A form of nat-20(S)-OHC with a terminal alkyne group, which can be used in linking reactions known as click chemistry; has been conjugated with magnetic beads to demonstrate that nat-20(S)-OHC directly binds SMO
Catalog No. 50-289-3987