Learn More
Medchemexpress LLC Natalizumab | 189261-10-7 | ≥99.0% | 146.22 kDa | 5 MG
SDP
Supplier:  Medchemexpress LLC HY108831A5MG
Encompass_Preferred
Natalizumab is a recombinant, humanized IgG4 monoclonal antibody that binds α4β1 integrin and prevents its interaction with vascular cell adhesion molecule-1. Supplied as a liquid solution for research use, the product is intended for studies of immune cell adhesion, migration, and inflammatory disease models; follow the datasheet for storage and handling and dilute or reconstitute with sterile PBS or saline as indicated.
  • Recombinant humanized IgG4 monoclonal antibody.
  • Binds α4β1 integrin and blocks VCAM-1 interaction.
  • Supplied as a liquid solution, ready for dilution or reconstitution.
  • Suitable for studies of immune cell adhesion, migration, and inflammatory disease models.
  • High purity (≥99.0%) and molecular weight approximately 146 kDa; consult the datasheet for storage and handling.
Catalog No. 50-002-63660