Learn More
Medchemexpress LLC Natamycin | 7681-93-8 | 98.5% | 1 ML

Supplier: Medchemexpress LLC HYB013310MM/1ML
Natamycin (Pimaricin) is a macrolide antibiotic agent derived from various Streptomyces strains. It inhibits fungal growth by preventing the transport of amino acids and glucose across the plasma membrane. It is used as a food preservative, an agricultural antifungal agent, and in research related to fungal keratitis. Unlike other polyene antibiotics, its mode of action does not rely on ergosterol-dependent permeabilization of the plasma membrane.
- Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
- In vitro solubility in DMSO: 15.38 mg/mL (23.10 mM)
- In vitro solubility in H₂O: Insoluble (< 0.1 mg/mL)
- In vivo solubility: ≥ 1.67 mg/mL (2.51 mM) in 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline (Clear solution)
- Alternative in vivo solubility: ≥ 1.67 mg/mL (2.51 mM) in 10% DMSO, 90% (20% SBE-β-CD in saline) (Clear solution)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.