Learn More
Selleck Chemical LLC Natamycin 200mg 7681-93-8 Pimaricin
SDP
Supplier:  Selleck Chemical LLC S1517200MG
Encompass_Preferred
Natamycin (Pimaricin), a natural and versatile anti-fungal agent during fermentation by the bacterium Streptomyces natalensis, commonly found in soil; with little to no flavour interference *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-791-5