Learn More
Abcam Native Human IgG4 protein, 1MG
Supplier:  Abcam AB1832661MG
Encompass_Preferred
Native Human IgG4 protein is a Human Full Length Immunoglobulin G4 (IgG4) protein with >95% purity and suitable for SDS-PAGE. The predicted molecular weight of ab183266 native protein is 146 kDa. -Save time and ensure accurate results - use ab183266 as an IgG4 isotype control
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant gamma 4, Ig gamma-4 chain C region, IGHG4
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1785074