Learn More
Abcam Native human Neutrophil Elastase protein (Active), 100UG
Supplier:  Abcam AB280938100UG
Encompass_Preferred
Native human Neutrophil Elastase protein (Active) is a Human Full Length protein, in the 30 to 267 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS, ELISA.
Protein/ Peptide/ Enzyme Synonyms: ELA2, ELANE, Neutrophil elastase, Bone marrow serine protease, Elastase-2, Human leukocyte elastase, Medullasin, PMN elastase, HLE
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4020