Learn More
Abcam Native Mouse Albumin protein (Azide free), 100MG
Supplier:  Abcam AB183228100MG
Encompass_Preferred
Native Mouse Albumin protein (Azide free) is a Mouse Full Length protein, in the 25 to 608 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Alb-1, Alb1, Alb, Albumin
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2386067