Learn More
Medchemexpress LLC Natriuretic Peptides A Antibody (YA1381) | 50 UL

Supplier: Medchemexpress LLC HYP8163650UL
Natriuretic Peptides A Antibody (YA1381) is a rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Natriuretic Peptides A. This antibody is stable under recommended storage conditions and is intended for research use only.
- Rabbit-derived and non-conjugated IgG monoclonal antibody
- Targets natriuretic peptides A
- Reactivity: human
- Application: WB, IP
- Molecular weight: 16 kDa
- Concentration: 0.35 mg/mL
- Formulation: 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% glycerol, 0.01% sodium azide and 0.05% BSA
- Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
- Shipped with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.