Learn More
Hampton Research NATRIX 2 NO12 185ML
SDP
Supplier:  Hampton Research HR291712
Encompass
HR2-917-12/Natrix 2 #12 (E12), 185 ml - 0.02 M Magnesium sulfate hydrate, 0.002 M Cobalt(II) chloride hexahydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 25% v/v (+/-)-2-Methyl-2,4-pentanediol, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-7