Learn More
Hampton Research NATRIX NO06 185ML
SDP
Supplier:  Hampton Research HR291806
Encompass
HR2-918-06/Natrix #06 (A6), 185 ml - 0.1 M Ammonium sulfate, 0.01 M Magnesium chloride hexahydrate, 0.05 M MES monohydrate pH 5.6, 20 % w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-170-9