Learn More
Medchemexpress LLC HY-P2373 5mg Medchemexpress, Nattokinase CAS:133876-92-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP23735MG
Encompass_Preferred
Medchemexpress, HY-P2373 5mg Nattokinase CAS:133876-92-3 Nattokinase is a potent fibrinolytic enzyme. Nattokinase can break down blood clots by directly hydrolyzing fibrin and plasmin substrate. Nattokinase can be used for the research of cardiovascular diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1722