Learn More
Medchemexpress LLC NaV1.7 inhibitor-1 | 1494585-79-3 | 99.2% | 50 MG

Supplier: Medchemexpress LLC HY11993450MG
NaV1.7 inhibitor-1 is a potent, selective inhibitor of the voltage-gated sodium channel NaV1.7 (IC50 0.6 nM) used in research on pain and sodium channel pharmacology. It is supplied as a white to off-white solid with high purity and is soluble in DMSO for in vitro studies.
- Potent NaV1.7 inhibition with an IC50 of 0.6 nM.
- Approximately 80-fold selectivity versus hNaV1.5.
- High purity suitable for research (99.17%).
- Soluble in DMSO (100 mg/mL) for in vitro applications.
- Powder stability with recommended storage at -20°C for long-term use.
- Available in multiple pack sizes including 50 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.