Learn More
TARGETMOL CHEMICALS INC Navitoclax-piperazine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T121861MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Navitoclax-piperazine (ABT-263-piperazine) is a B-cell lymphoma extra large (BCL-XL) inhibitor. Purity 99.11%
Catalog No. 50-287-8348